CAS RN 641-74-7
IUPAC Navn:
hexahydrofuro[3,2-b]furan-3,6-diol
Synonymer:
isosorbide
Molekylvægt (g/mol):
146.14
Molekylformel:
C6H10O4
InChi Key:
KLDXJTOLSGUMSJ-UHFFFAOYNA-N
SMILES:
OC1COC2C(O)COC12
1
–
3
af
3
Resultater
1
1,4:3,6-Dianhydro-D-mannitol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,4:3,6-Dianhydro-D-mannitol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,4:3,6-Dianhydro-D-mannitol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.