CAS RN 631920-67-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
1-C-[2,4-Bis(1,1-dimethylethoxy)-5-pyrimidinyl]-5-O-(1-methoxy-1-methylethyl)-2,3-O-(1-methylethylidene)-Alpha-D-ribofuranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-C-[2,4-Bis(1,1-dimethylethoxy)-5-pyrimidinyl]-5-O-(1-methoxy-1-methylethyl)-2,3-O-(1-methylethylidene)-Alpha-D-ribofuranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.