CAS RN 6232-57-1
IUPAC Navn:
4-[2-(4-aminophenyl)diazen-1-yl]-2-methoxy-5-methylaniline
Synonymer:
4-[2-(4-aminophenyl)diazen-1-yl]-2-methoxy-5-methylaniline
Molekylvægt (g/mol):
256.31
Molekylformel:
C14H16N4O
InChi Key:
DBENZRUFCCLWPP-UHFFFAOYSA-N
SMILES:
COC1=CC(N=NC2=CC=C(N)C=C2)=C(C)C=C1N
1
–
3
af
3
Resultater
1
Disperse Diazo Black 3BF, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Disperse Diazo Black 3BF, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Disperse Diazo Black 3BF, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.