CAS RN 621-87-4
IUPAC Navn:
1-phenoxypropan-2-one
Synonymer:
2-propanone, 1-phenoxy-
Molekylvægt (g/mol):
150.18
Molekylformel:
C9H10O2
InChi Key:
QWAVNXZAQASOML-UHFFFAOYSA-N
SMILES:
CC(=O)COC1=CC=CC=C1
1
–
3
af
3
Resultater
1
Phenoxy-2-propanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Phenoxy-2-propanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Phenoxy-2-propanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.