CAS RN 61848-66-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
[SP-4-2-(1R-trans)]-(1,2-Cyclohexanediamine-N,N’) Dichloridoplatinum(II) >85%, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
[SP-4-2-(1R-trans)]-(1,2-Cyclohexanediamine-N,N') Dichloridoplatinum(II) >85%, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.