CAS RN 60456-22-6
IUPAC Navn:
(4R)-4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane
Synonymer:
(4R)-4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane
Molekylvægt (g/mol):
150.6
Molekylformel:
C6H11ClO2
InChi Key:
BNPOTXLWPZOESZ-YFKPBYRVSA-N
SMILES:
CC1(C)OC[C@H](CCl)O1
1
–
4
af
4
Resultater
1
(S)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(S)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(S)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.