CAS RN 6027-89-0
IUPAC Navn:
(2S,3S,4R,5S)-2,3,4,5,6-pentahydroxyhexanal
Synonymer:
L-gulose aldehydo-L-gulose
Molekylvægt (g/mol):
180.16
Molekylformel:
C6H12O6
InChi Key:
GZCGUPFRVQAUEE-JGWLITMVSA-N
SMILES:
OC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)C=O
1
–
2
af
2
Resultater
1
L-Gulose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
L-Gulose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.