CAS RN 593274-97-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Integrin Antagonists 27, MedChemExpress
MedChemExpress Integrin Antagonists 27 is a small molecule integrin (alpha)v(beta)3 antagonist with binding affinity of 18 nM, as s novel anticancer agent.