CAS RN 579-10-2
IUPAC Navn:
N-methyl-N-phenylacetamide
Synonymer:
N-methylacetanilide
Molekylvægt (g/mol):
149.19
Molekylformel:
C9H11NO
InChi Key:
LMTGCJANOQOGPI-UHFFFAOYSA-N
SMILES:
CN(C(C)=O)C1=CC=CC=C1
1
–
1
af
1
Resultater
1
N-Methylacetanilide, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality