CAS RN 574749-31-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(2R,3R)-2-[3,4-Bis(phenylmethoxy)phenyl]-2,3-dihydro-3,5,7-tris(phenylmethoxy)-4H-1-benzopyran-4-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2R,3R)-2-[3,4-Bis(phenylmethoxy)phenyl]-2,3-dihydro-3,5,7-tris(phenylmethoxy)-4H-1-benzopyran-4-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.