CAS RN 55954-23-9
IUPAC Navn:
methyl 2-(2,4-dichlorophenyl)acetate
Synonymer:
methyl 2-(2,4-dichlorophenyl)acetate
Molekylvægt (g/mol):
219.06
Molekylformel:
C9H8Cl2O2
InChi Key:
SRCVNBFTEARRJY-UHFFFAOYSA-N
SMILES:
COC(=O)CC1=CC=C(Cl)C=C1Cl
1
–
2
af
2
Resultater
1
DCAA Methyl Ester 100 μg/mL in Methyl tert-butyl ether, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality