CAS RN 556828-95-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Methyl (2Z)-2-(acetylamino)-3-[1,1′-biphenyl]-4-yl-2-propenoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methyl (2Z)-2-(acetylamino)-3-[1,1′-biphenyl]-4-yl-2-propenoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Methyl (2Z)-2-(acetylamino)-3-[1,1′-biphenyl]-4-yl-2-propenoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.