CAS RN 548783-71-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Fexofenadine-D6, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Fexofenadine-d6, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Fexofenadine-d6, MedChemExpress
MedChemExpress Fexofenadine D6 (MDL-16455 D6) is deuterium labeled is Fexofenadine, which is an antihistamine pharmaceutical agent.