CAS RN 5423-23-4
IUPAC Navn:
bis(guanidine); phosphoric acid
Synonymer:
bis(guanidine); phosphoric acid
Molekylvægt (g/mol):
216.14
Molekylformel:
C2H13N6O4P
InChi Key:
SSHPJTCTVWVCLI-UHFFFAOYSA-N
SMILES:
NC(N)=N.NC(N)=N.OP(O)(O)=O
1
–
3
af
3
Resultater
1
Guanidine Phosphate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Guanidine Phosphate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Guanidine Phosphate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.