CAS RN 52818-63-0
IUPAC Navn:
N-[(4-methoxyphenyl)methyl]pyridin-2-amine
Synonymer:
N-[(4-methoxyphenyl)methyl]pyridin-2-amine
Molekylvægt (g/mol):
214.27
Molekylformel:
C13H14N2O
InChi Key:
SMJGJENXFSWIMQ-UHFFFAOYSA-N
SMILES:
COC1=CC=C(CNC2=CC=CC=N2)C=C1
1
–
1
af
1
Resultater
1
Mepyramine Maleate EP Impurity A, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.