CAS RN 52029-86-4
IUPAC Navn:
11-oxo-3,10-diazapentacyclo[10.7.1.0²,¹⁰.0⁴,⁹.0¹⁶,²⁰]icosa-1(20),2,4,6,8,12,14,16,18-nonaene-17-carboxylic acid
Synonymer:
11-oxo-3,10-diazapentacyclo[10.7.1.0²,¹⁰.0⁴,⁹.0¹⁶,²⁰]icosa-1(20),2,4,6,8,12,14,16,18-nonaene-17-carboxylic acid
Molekylvægt (g/mol):
314.3
Molekylformel:
C19H10N2O3
InChi Key:
MYKOWOGZBMOVBJ-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C2C=CC=C3C(=O)N4C5=CC=CC=C5N=C4C(C=C1)=C23
1
–
1
af
1
Resultater
1
STO-609, MedChemExpress
MedChemExpress STO-609 is a selective and cell-permeable inhibitor of the Ca2+/calmodulin-dependent protein kinase kinase (CaM-KK), with Ki values of 80 and 15 ng/mL for recombinant CaM-KKα and CaM-KKβ, respectively. STO-609 inhibits AMP-activated protein kinase kinase (AMPKK) activity in HeLa cell lysates with an IC50 ∼0.02 g/ml.