CAS RN 51591-75-4
IUPAC Navn:
1,3-dibenzyl-2-oxoimidazolidine-4,5-dicarboxylic acid
Synonymer:
1,3-dibenzyl-2-oxoimidazolidine-4,5-dicarboxylic acid
Molekylvægt (g/mol):
354.36
Molekylformel:
C19H18N2O5
InChi Key:
QSMUFXXTSUEZJA-UHFFFAOYNA-N
SMILES:
OC(=O)C1C(N(CC2=CC=CC=C2)C(=O)N1CC1=CC=CC=C1)C(O)=O
1
–
3
af
3
Resultater
1
cis-1,3-Dibenzyl-2-imidazolidone-4,5-dicarboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
cis-1,3-Dibenzyl-2-imidazolidone-4,5-dicarboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
cis-1,3-Dibenzyl-2-imidazolidone-4,5-dicarboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.