CAS RN 506-52-5
IUPAC Navn:
hexacosan-1-ol
Synonymer:
ceryl alcohol cerylic alcohol
Molekylvægt (g/mol):
382.72
Molekylformel:
C26H54O
InChi Key:
IRHTZOCLLONTOC-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCCCCCCCCCO
1
–
2
af
2
Resultater
1
1-Hexacosanol (>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-Hexacosanol (>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.