CAS RN 503559-84-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Glucagon receptor antagonists-1, MedChemExpress
MedChemExpress Glucagon receptor antagonists-1 is a highly potent glucagon receptor antagonist.