CAS RN 501120-40-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
5-(2-Ethoxyphenyl)-2,6-dihydro-2-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-(2-Ethoxyphenyl)-2,6-dihydro-2-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.