CAS RN 50-44-2
IUPAC Navn:
6,7-dihydro-3H-purine-6-thione
Synonymer:
mercaptopurine Mern leupurin ismipur mercaleukin mercaleukim 6-mercaptopurine purimethol mercaptopurine monohydrate leukerin
Molekylvægt (g/mol):
152.18
Molekylformel:
C5H4N4S
InChi Key:
GLVAUDGFNGKCSF-UHFFFAOYSA-N
SMILES:
S=C1N=CNC2=C1NC=N2
1
–
1
af
1
Resultater
1
6-Mercaptopurine, MedChemExpress
MedChemExpress 6-Mercaptopurine is a purine analogue which acts as an antagonist of the endogenous purines and has been widely used as antileukemic agent and immunosuppressive drug.