CAS RN 488832-69-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Elesclomol, MedChemExpress
MedChemExpress Elesclomol (STA-4783) is a potent copper ionophore and promotes copper-dependent cell death (cuproptosis).