CAS RN 479065-02-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
3-[(1RS)-5-Bromo-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-1-yl]-N,N-dimethylpropan-1-amine Hydrobromide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.