CAS RN 475502-13-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
2-Acetamido-4-O-(2-acetamido-2-deoxy-Beta-D-glucopyranosyl)-1,6-di-O-N-acetyl-Alpha-D-muramic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.