CAS RN 4477-79-6
IUPAC Navn:
(1Z)-1-(2-{2,5-dimethyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-1,2-dihydronaphthalen-2-one
Synonymer:
solvent red 26
Molekylvægt (g/mol):
394.48
Molekylformel:
C25H22N4O
InChi Key:
OLDHUFONGVQOBH-ZOQJIKCCSA-N
SMILES:
CC1=CC=CC=C1N=NC1=C(C)C=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C(C)=C1
1
–
1
af
1
Resultater
1
Solvent Red 26, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality