CAS RN 4386-25-8
IUPAC Navn:
5-chloro-2-hydroxy-3-{2-[(1Z)-2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene]hydrazin-1-yl}benzene-1-sulfonic acid
Synonymer:
5-chloro-2-hydroxy-3-{2-[(1Z)-2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene]hydrazin-1-yl}benzenesulfonic acid
Molekylvægt (g/mol):
344.72
Molekylformel:
C12H9ClN2O6S
InChi Key:
RQIJHLQABPWAQV-ZSOIEALJSA-N
SMILES:
OC1=CC(=O)C=C\C1=N\NC1=CC(Cl)=CC(=C1O)S(O)(=O)=O
1
–
1
af
1
Resultater
1
Lumogallion, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.