CAS RN 438244-32-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
9,11-Dehydro (17a)-Trimethyl(prop-1-yn-1-yl)silyl Estradiol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
9,11-Dehydro (17Alpha)-Trimethyl(prop-1-yn-1-yl)silyl Estradiol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
9,11-Dehydro (17Alpha)-Trimethyl(prop-1-yn-1-yl)silyl Estradiol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.