CAS RN 4137-56-8
IUPAC Navn:
{6-methoxy-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl}methyl 4-methylbenzene-1-sulfonate
Synonymer:
{6-methoxy-2,2-dimethyl-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl}methyl 4-methylbenzenesulfonate
Molekylvægt (g/mol):
358.41
Molekylformel:
C16H22O7S
InChi Key:
IAPMZKRZMYQZSW-UHFFFAOYNA-N
SMILES:
COC1OC(COS(=O)(=O)C2=CC=C(C)C=C2)C2OC(C)(C)OC12
1
–
2
af
2
Resultater
1
Methyl 2,3-O-Isopropylidene-5-O-p-tolylsulfonyl-ß-D-ribofuranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methyl 2,3-O-Isopropylidene-5-O-p-tolylsulfonyl-ß-D-ribofuranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.