CAS RN 402-40-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
1,5-Bis(4-Allyldimethylammoniumphenyl)pentan-3-one, Dibromide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,5-Bis(4-Allyldimethylammoniumphenyl)pentan-3-one, Dibromide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.