CAS RN 3967-32-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
5-(1-Methyl-4-piperidinyl)-5H-dibenzo[a,d]cyclohepten-5-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-(1-Methyl-4-piperidinyl)-5H-dibenzo[a,d]cyclohepten-5-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5-(1-Methyl-4-piperidinyl)-5H-dibenzo[a,d]cyclohepten-5-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.