CAS RN 364071-16-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
OT-R antagonist 2, MedChemExpress
MedChemExpress OT-R antagonist 2 is a nonpeptide low molecular weight OT-R antagonist.