CAS RN 35693-92-6
IUPAC Navn:
2,4,6-trichloro-1,1'-biphenyl
Synonymer:
2,4,6-trichlorobiphenyl
Molekylvægt (g/mol):
257.54
Molekylformel:
C12H7Cl3
InChi Key:
MTLMVEWEYZFYTH-UHFFFAOYSA-N
SMILES:
ClC1=CC(Cl)=C(C(Cl)=C1)C1=CC=CC=C1
1
–
3
af
3
Resultater
3
PCB No. 30 100 μg/mL in Hexane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality