CAS RN 35607-20-6
IUPAC Navn:
2-{[3-(dioctadecylamino)propyl](2-hydroxyethyl)amino}ethan-1-ol
Synonymer:
avridine avridina avridine phosphate (2:1) avridinum
Molekylvægt (g/mol):
667.21
Molekylformel:
C43H90N2O2
InChi Key:
WXNRAKRZUCLRBP-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCN(CCCCCCCCCCCCCCCCCC)CCCN(CCO)CCO
1
–
2
af
2
Resultater
1
Avridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Avridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.