CAS RN 352-97-6
IUPAC Navn:
2-[(diaminomethylidene)amino]acetic acid
Synonymer:
β-guanidinoacetic acid
Molekylvægt (g/mol):
117.11
Molekylformel:
C3H7N3O2
InChi Key:
BPMFZUMJYQTVII-UHFFFAOYSA-N
SMILES:
NC(N)=NCC(O)=O
1
–
2
af
2
Resultater
1
Guanidinoacetic acid, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality