CAS RN 3513-03-9
IUPAC Navn:
6-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3-[3-amino-5-(N-methylcarbamimidamido)pentanamido]-3,6-dihydro-2H-pyran-2-carboxylic acid hydrochloride
Synonymer:
blasticidin-S hydrochloride
Molekylvægt (g/mol):
458.9
Molekylformel:
C17H27ClN8O5
InChi Key:
YQXYQOXRCNEATG-UHFFFAOYNA-N
SMILES:
Cl.CN(CCC(N)CC(=O)NC1C=CC(OC1C(O)=O)N1C=CC(N)=NC1=O)C(N)=N
1
–
3
af
3
Resultater
1
Blasticidin S Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Blasticidin S Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Blasticidin S Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.