CAS RN 342002-82-8
CAS RN 342002-82-8
IUPAC Navn:
{4-[(propan-2-yloxy)carbonyl]phenyl}boronic acid
Synonymer:
4-(isopropoxycarbonyl)phenylboronic acid
Molekylvægt (g/mol):
208.02
Molekylformel:
C10H13BO4
InChi Key:
NIQHFKWRHJZMQU-UHFFFAOYSA-N
SMILES:
CC(C)OC(=O)C1=CC=C(C=C1)B(O)O
1
–
1
af
1
Resultater
1
4-(Isopropoxycarbonyl)benzeneboronic acid, 98%, Thermo Scientific™
CAS: 342002-82-8 Molekylær formel: C10H13BO4 Molekylvægt (g/mol): 208.02 MDL nummer: MFCD02259385 InChI nøgle: NIQHFKWRHJZMQU-UHFFFAOYSA-N Synonym: 4-isopropoxycarbonylphenylboronic acid,4-isopropoxycarbonyl phenyl boronic acid,4-isopropoxycarbonyl benzeneboronic acid,propan-2-yl 4-dihydroxyboranyl benzoate,isopropyl 4-boronobenzoate,4-isopropoxycarbonyl phenylboronic acid,p-isopropoxycarbonyl phenylboronic acid,4-isopropoxycarbonylphenylboronicacid,4-propan-2-yloxycarbonylphenyl boronic acid,acmc-209i5o PubChem CID: 2734703 IUPAC navn: (4-propan-2-yloxycarbonylphenyl)boronic acid SMIL: CC(C)OC(=O)C1=CC=C(C=C1)B(O)O