CAS RN 34193-38-9
IUPAC Navn:
butan-1-ol
Synonymer:
butanol n butyl alcohol butylowy alkohol fema number 2178 butan-1-ol (substance) N-butanol butanolo butyric or normal primary butyl alcohol alcohol,butyl normal primary butyl alcohol
Molekylvægt (g/mol):
74.12
Molekylformel:
C4H10O
InChi Key:
LRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILES:
CCCCO
1
–
2
af
2
Resultater
1
1-Butanol-d10 (n-Butyl Alcohol-d10), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
n-Butyl Alcohol-d10, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.