CAS RN 3290-92-4
IUPAC Navn:
2,2-bis({[(2-methylprop-2-enoyl)oxy]methyl})butyl 2-methylprop-2-enoate
Synonymer:
TMPT light ester tmp tmpt (crosslinking agent) acryester tmp chemlink 30 sartomer 350 NK ester tmpt saret 515 PTMA tmptma
Molekylvægt (g/mol):
338.4
Molekylformel:
C18H26O6
InChi Key:
OKKRPWIIYQTPQF-UHFFFAOYSA-N
SMILES:
CCC(COC(=O)C(C)=C)(COC(=O)C(C)=C)COC(=O)C(C)=C
1
–
1
af
1
Resultater
1
1,1,1-Trimethylolpropane trimethacrylate 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality