CAS RN 3031-66-1
IUPAC Navn:
hex-3-yne-2,5-diol
Synonymer:
3-hexyne-2,5-diol
Molekylvægt (g/mol):
114.14
Molekylformel:
C6H10O2
InChi Key:
KDOWHHULNTXTNS-UHFFFAOYNA-N
SMILES:
CC(O)C#CC(C)O
1
–
1
af
1
Resultater
1
3-Hexyne-2,5-diol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.