CAS RN 303-43-5
IUPAC Navn:
(1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl (9Z)-octadec-9-enoate
Synonymer:
cholesteryl oleate cholesteryl oleate, (e)-isomer cholesteryl oleate, (e)- 5-cholesten-3b-ol 3-oleate cholesteryl [9,10-3H]oleate cholest-5-en-3b-yl cholesteryl oleate, (E)- (e)-cholesteryl oleate cholesteryl oleate, (E)-isomer cholesteroyl-oleate
Molekylvægt (g/mol):
651.12
Molekylformel:
C45H78O2
InChi Key:
RJECHNNFRHZQKU-RMUVNZEASA-N
SMILES:
CCCCCCCC\C=C/CCCCCCCC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC=C2C1)[C@H](C)CCCC(C)C
1
–
2
af
2
Resultater
1
Cholesteryl Oleate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Cholesteryl Oleate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.