CAS RN 302-23-8
IUPAC Navn:
1-acetyl-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl acetate
Synonymer:
hydroxyprogesterone acetate
Molekylvægt (g/mol):
372.51
Molekylformel:
C23H32O4
InChi Key:
VTHUYJIXSMGYOQ-UHFFFAOYNA-N
SMILES:
CC(=O)OC1(CCC2C3CCC4=CC(=O)CCC4(C)C3CCC12C)C(C)=O
1
–
2
af
2
Resultater
1
Progesterone Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3,20-Dioxopregn-4-en-17-yl Acetate (Hydroxyprogesterone Acetate), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.