CAS RN 3016286-99-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Kampagner er tilgængelige
Perfluoroundecanesulfonic acid sodium 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Perfluoroundecanesulfonic acid sodium 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality