CAS RN 3007-97-4
IUPAC Navn:
ethyl naphthalene-1-carboxylate
Synonymer:
ethyl naphthalene-1-carboxylate
Molekylvægt (g/mol):
200.24
Molekylformel:
C13H12O2
InChi Key:
XCTLDQQOHIEUCJ-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C2C=CC=CC2=CC=C1
1
–
1
af
1
Resultater
1
Ethyl 1-naphthoate, 97%
CAS: 3007-97-4 Molekylær formel: C13H12O2 Molekylvægt (g/mol): 200.237 MDL nummer: MFCD00016805 InChI nøgle: XCTLDQQOHIEUCJ-UHFFFAOYSA-N Synonym: ethyl 1-naphthoate,ethyl alpha-naphthoate,1-naphthoic acid, ethyl ester,ethyl alfa-naphthoate,1-naphthalenecarboxylic acid, ethyl ester,1-naphthoic acid ethyl ester,naphthalene-1-carboxylic acid ethyl ester,ethyl naphthalenecarboxylate,ethyl a-naphthoate,ethylalpha-naphthoate PubChem CID: 76364 IUPAC navn: ethylnaphthalen-1-carboxylat SMIL: CCOC(=O)C1=CC=CC2=CC=CC=C21