CAS RN 2905-54-6
IUPAC Navn:
methyl 2,3-dichlorobenzoate
Synonymer:
methyl 2,3-dichlorobenzoate
Molekylvægt (g/mol):
205.03
Molekylformel:
C8H6Cl2O2
InChi Key:
RFVULUSGHFRDHJ-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=CC(Cl)=C1Cl
1
–
1
af
1
Resultater
1
Methyl 2,3-dichlorobenzoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.