CAS RN 2899-36-7
IUPAC Navn:
hydrogen ethyl 2-amino-4-(methylsulfanyl)butanoate chloride
Synonymer:
hydrogen ethyl 2-amino-4-(methylsulfanyl)butanoate chloride
Molekylvægt (g/mol):
213.72
Molekylformel:
C7H16ClNO2S
InChi Key:
KPWCQEUBJAIORR-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CCOC(=O)C(N)CCSC
1
–
1
af
1
Resultater
1
L-Methionine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals
CAS: 2899-36-7 Molekylær formel: C7H15NO2S·HCl Molekylvægt (g/mol): 213.73 InChI nøgle: KPWCQEUBJAIORR-RGMNGODLSA-N Synonym: l-methionine ethyl ester hydrochloride,ethyl l-methionate hydrochloride,h-met-oet.hcl,ethyll-methionatehydrochloride,l-methionine, ethyl ester, hydrochloride,h-met-oet hcl,ethyl l-methionate hcl,ethyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride,h-met-oet?cl,pubchem14924 PubChem CID: 3083785 IUPAC navn: ethyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride SMIL: CCOC(=O)C(CCSC)N.Cl