CAS RN 2733558-78-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
11-Cyclopropyl-5,11-dihydro-4-(hydroxymethyl)-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one-d2, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
11-Cyclopropyl-5,11-dihydro-4-(hydroxymethyl)-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one-d2, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
11-Cyclopropyl-5,11-dihydro-4-(hydroxymethyl)-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one-d2, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.