CAS RN 24353-61-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Isocarbophos, TRC
CAS: 24353-61-5 Molekylær formel: C11 H16 N O4 P S Molekylvægt (g/mol): 289.29 Synonym: Salicylic acid, isopropyl ester, O-ester with O-methyl phosphoramidothioate (8CI),Phosphoramidothioic acid, O-methyl ester, O-ester with isopropyl salicylate (8CI),BAY 93820,Isocarbofos,Isocarbophos,Isopropyl O-(methoxyaminothiophosphoryl) salicylate,Optunal IUPAC navn: propan-2-yl 2-[amino(methoxy)phosphinothioyl]oxybenzoate SMIL: COP(=S)(N)Oc1ccccc1C(=O)OC(C)C