CAS RN 23930-19-0
IUPAC Navn:
(1S,3aS,3bS,5aS,7R,9aS,9bS,11aS)-1-acetyl-7-hydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-10-one
Synonymer:
alfaxalone
Molekylvægt (g/mol):
332.48
Molekylformel:
C21H32O3
InChi Key:
DUHUCHOQIDJXAT-OLVMNOGESA-N
SMILES:
CC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4C[C@H](O)CC[C@]4(C)[C@H]3C(=O)C[C@]12C
1
–
2
af
2
Resultater
1
Alphaxalone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Alphaxalone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.