CAS RN 2376841-30-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ HM Janelia Fluor 526, SE
Fluorogenic spontaneously blinking yellow-emitting dye (Reactive group: NHS ester). Application: dSTORM, SmLSM, STED, SIM and confocal microscopy