CAS RN 2357110-58-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ Thalidomide 5'-amine-PEG2-amine
Cereblon ligand with PEG linker and terminal amine for onward chemistry